siRNA Details
| Gene Symbol: Myrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033481.1 | NP_001028653.1 | 25 | 3350 | ||
| GenBank mRNA | AK039649.1 | - | 16 | 1712 | ||
| AK141695.1 | - | 24 | 3601 | |||
| BC157942.1 | - | 23 | 2822 | 9007277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myrf| Gene Symbol: Myrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033481.1 | NP_001028653.1 | 25 | 3350 | ||
| GenBank mRNA | AK039649.1 | - | 16 | 1712 | ||
| AK141695.1 | - | 24 | 3601 | |||
| BC157942.1 | - | 23 | 2822 | 9007277 | ||