siRNA Details
| Gene Symbol: Xpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009531.2 | NP_033557.2 | 9 | 1545 | ||
| GenBank mRNA | AB071144.1 | - | 9 | 1482 | ||
| AK004713.1 | - | 9 | 1485 | |||
| AK028595.1 | - | 9 | 1483 | |||
| AK166981.1 | - | 9 | 1480 | |||
| AK170975.1 | - | 9 | 1477 | |||
| U27398.1 | - | 9 | 1456 | |||
| U40005.1 | - | 8 | 1284 | |||

Xpc
Genomic Map