siRNA Details
| Gene Symbol: D030056L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177640.4 | NP_808308.1 | 1, 2 | 421 | ||
| GenBank mRNA | AK083631.1 | - | 1, 2 | 422 | ||
| AK155389.1 | - | 1, 2 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D030056L22Rik| Gene Symbol: D030056L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177640.4 | NP_808308.1 | 1, 2 | 421 | ||
| GenBank mRNA | AK083631.1 | - | 1, 2 | 422 | ||
| AK155389.1 | - | 1, 2 | 462 | |||