siRNA Details
Gene Symbol: D030056L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177640.4 | NP_808308.1 | 2 | 447 | ||
GenBank mRNA | AK083631.1 | - | 2 | 448 | ||
AK145019.1 | - | 2 | 495 | |||
AK155389.1 | - | 2 | 488 | |||
AK213250.1 | - | 1 | 55 | |||
BC020125.1 | - | 2 | 428 | 3985075 |
