siRNA Details
| Gene Symbol: Ermp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081213.1 | NP_001074682.1 | 4 | 857 | ||
| GenBank mRNA | AK041066.1 | - | 4 | 828 | ||
| AK050284.1 | - | 4 | 853 | |||
| AK081674.1 | - | 3 | 374 | |||
| AK129447.1 | - | 4 | 785 | |||
| AK137206.1 | - | 4 | 813 | |||
| BC086627.1 | - | 4 | 798 | |||
| BC151181.1 | - | 4 | 816 | 9088070 | ||
| BC151182.1 | - | 4 | 816 | 9088071 | ||
| BC157914.1 | - | 4 | 816 | 9088152 | ||

Ermp1
Genomic Map