siRNA Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130525.1 | NP_001123997.1 | 3 | 1194 | ||
| NM_001130526.1 | NP_001123998.1 | 3 | 1186 | |||
| NM_145503.2 | NP_663478.2 | 3 | 1238 | |||
| GenBank mRNA | AK081367.1 | - | 3 | 1112 | ||
| AK129446.1 | - | 2 | 1103 | |||
| AK144154.1 | - | 3 | 1205 | |||
| AK169936.1 | - | 3 | 1227 | |||
| AK176990.1 | - | 1 | 16 | |||
| BC014695.1 | - | 2 | 1087 | 3157493 | ||
| BC092263.1 | - | 3 | 1218 | 2654420 | ||

Lzts2
Genomic Map