siRNA Details
| Gene Symbol: Prlhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC125649.1 | - | 1 | 465 | 40130164 | |
| BC125651.1 | - | 1 | 465 | 40130166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prlhr| Gene Symbol: Prlhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC125649.1 | - | 1 | 465 | 40130164 | |
| BC125651.1 | - | 1 | 465 | 40130166 | ||