siRNA Details
| Gene Symbol: Dyrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145508.2 | NP_663483.1 | 3 | 843 | ||
| GenBank mRNA | AK035114.1 | - | 622 | |||
| AK161495.1 | - | 3 | 755 | |||
| BC006704.1 | - | 3 | 843 | 3584001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dyrk3| Gene Symbol: Dyrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145508.2 | NP_663483.1 | 3 | 843 | ||
| GenBank mRNA | AK035114.1 | - | 622 | |||
| AK161495.1 | - | 3 | 755 | |||
| BC006704.1 | - | 3 | 843 | 3584001 | ||