siRNA Details
| Gene Symbol: Rab29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB232625.1 | - | 4 | 416 | ||
| BC016133.1 | - | 5 | 668 | 4485029 | ||
| BC029056.1 | - | 4 | 457 | 4947517 | ||
| CK796080.1 | - | 2 | 528 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab29| Gene Symbol: Rab29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB232625.1 | - | 4 | 416 | ||
| BC016133.1 | - | 5 | 668 | 4485029 | ||
| BC029056.1 | - | 4 | 457 | 4947517 | ||
| CK796080.1 | - | 2 | 528 | |||