siRNA Details
| Gene Symbol: Ipo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153774.1 | NP_722469.1 | 4 | 505 | ||
| GenBank mRNA | AF273672.1 | - | 4 | 450 | ||
| AF273673.1 | - | 4 | 450 | |||
| AJ309238.1 | - | 4 | 444 | |||
| AK036687.1 | - | 4 | 455 | |||
| AK173127.1 | - | 4 | 445 | |||
| BC056469.1 | - | 4 | 472 | |||
| BC079546.1 | - | 5 | 616 | |||
| BC098508.1 | - | 4 | 455 | 30015513 | ||
| CD347972.1 | - | 5 | 616 | |||

Ipo9
Genomic Map