siRNA Details
Gene Symbol: BC003331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077237.1 | NP_001070705.1 | 7 | 959 | ||
NM_145511.2 | NP_663486.2 | 7 | 959 | |||
GenBank mRNA | AK152022.1 | - | 5 | 475 | ||
AK152955.1 | - | 5 | 475 | |||
BC003331.1 | - | 6 | 614 | 3256634 | ||
CB235715.1 | - | 7 | 657 | |||
CX243768.1 | - | 3 | 207 | |||
DQ077804.2 | - | 6 | 573 |
