siRNA Details
| Gene Symbol: Rasal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177644.5 | NP_808312.3 | 6, 7 | 1492 | ||
| GenBank mRNA | AK039584.1 | - | 6, 7 | 1492 | ||
| AK086812.1 | - | 6, 7 | 1118 | |||
| BC120902.2 | - | 6, 7 | 835 | 40086771 | ||

Rasal2
Genomic Map