siRNA Details
| Gene Symbol: BC026585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033284.1 | NP_001028456.1 | 9 | 842 | ||
| GenBank mRNA | AK143913.1 | - | 9 | 842 | ||
| BC026585.1 | - | 9 | 844 | |||
| BC045198.1 | - | 9 | 839 | 5065108 | ||
| BC057972.1 | - | 3 | 344 | |||

BC026585
Genomic Map