siRNA Details
| Gene Symbol: Fmo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178038.1 | NP_001171509.1 | 2 | 354 | ||
| GenBank mRNA | BC051402.1 | - | 3 | 370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo6| Gene Symbol: Fmo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178038.1 | NP_001171509.1 | 2 | 354 | ||
| GenBank mRNA | BC051402.1 | - | 3 | 370 | ||