siRNA Details
Gene Symbol: Gm4847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164312.1 | NP_001157784.1 | 6, 7 | 1038 | ||
GenBank mRNA | AK146137.1 | - | 6, 7 | 1041 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164312.1 | NP_001157784.1 | 6, 7 | 1038 | ||
GenBank mRNA | AK146137.1 | - | 6, 7 | 1041 |