siRNA Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172650.2 | NP_766238.1 | 3, 4 | 405 | ||
GenBank mRNA | AK033491.1 | - | 3, 4 | 406 | ||
AK034380.1 | - | 3, 4 | 405 | |||
AK049362.1 | - | 3, 4 | 403 | |||
AK087865.1 | - | 3, 4 | 406 | |||
AK139700.1 | - | 3, 4 | 405 | |||
BC061458.1 | - | 3, 4 | 393 | |||
BC079581.1 | - | 3, 4 | 392 | 5702630 |
