siRNA Details
| Gene Symbol: Zfp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037707.1 | NP_001032796.1 | 5 | 913 | ||
| NM_001285797.1 | NP_001272726.1 | 5 | 1185 | |||
| NM_001285798.1 | NP_001272727.1 | 5 | 1139 | |||
| NM_011754.2 | NP_035884.2 | 5 | 908 | |||
| GenBank mRNA | AK028500.1 | - | 5 | 1140 | ||
| AK135293.1 | - | 5 | 1189 | |||
| AK147732.1 | - | 5 | 903 | |||
| BC059037.1 | - | 3 | 852 | 6812360 | ||

Zfp27
Genomic Map