siRNA Details
| Gene Symbol: Zfp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009556.3 | NP_033582.2 | 4 | 489 | ||
| GenBank mRNA | AB221555.1 | - | 1 | 88 | ||
| BC053399.1 | - | 4 | 462 | 30031349 | ||
| BC064785.1 | - | 5 | 521 | |||
| BC098467.1 | - | 4 | 424 | 30008039 | ||
| M28382.1 | - | 4 | 450 | |||

Zfp42
Genomic Map