siRNA Details
| Gene Symbol: Usp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033291.2 | NP_001028463.1 | 7 | 841 | ||
| NM_001198573.1 | NP_001185502.1 | 7 | 841 | |||
| GenBank mRNA | AK050239.1 | - | 7 | 838 | ||
| AK132295.1 | - | 7 | 841 | |||
| AK166303.1 | - | 7 | 818 | |||
| BC030843.1 | - | 7 | 790 | |||
| BC089376.1 | - | 4 | 343 | |||
| BC117957.1 | - | 8 | 956 | 40098174 | ||
| BC117958.1 | - | 7 | 824 | 40098175 | ||

Usp40
Genomic Map