siRNA Details
| Gene Symbol: C1ql3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153155.2 | NP_694795.1 | 2 | 694 | ||
| GenBank mRNA | AB044560.1 | - | 1577 | |||
| BC024634.1 | - | 2 | 694 | 4240136 | ||
| EU399230.1 | - | 2 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1ql3| Gene Symbol: C1ql3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153155.2 | NP_694795.1 | 2 | 694 | ||
| GenBank mRNA | AB044560.1 | - | 1577 | |||
| BC024634.1 | - | 2 | 694 | 4240136 | ||
| EU399230.1 | - | 2 | 677 | |||