siRNA Details
| Gene Symbol: Tmem203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177344.3 | NP_796318.2 | 1 | 432 | ||
| GenBank mRNA | AK050191.1 | - | 1 | 417 | ||
| AK051471.1 | - | 1 | 366 | |||
| AX775749.1 | - | 1 | 420 | |||
| BC022606.1 | - | 1 | 400 | 4165766 | ||
| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CD354207.1 | - | 149 | |||

Tmem203
Genomic Map