siRNA Details
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153125.2 | NP_694765.2 | 2 | 1174 | ||
GenBank mRNA | AA388295.1 | - | 1 | 198 | ||
CA463107.1 | - | 1 | 55 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153125.2 | NP_694765.2 | 2 | 1174 | ||
GenBank mRNA | AA388295.1 | - | 1 | 198 | ||
CA463107.1 | - | 1 | 55 |