siRNA Details
| Gene Symbol: Slc2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177627.1 | NP_001171098.1 | 9 | 1385 | ||
| NM_172659.2 | NP_766247.2 | 10 | 1547 | |||
| GenBank mRNA | AK079650.1 | - | 10 | 1492 | ||
| AK089246.1 | - | 10 | 1539 | |||
| AK137441.1 | - | 9 | 1762 | |||
| AK150104.1 | - | 10 | 1500 | |||
| AK166312.1 | - | 10 | 1505 | |||
| AK218156.1 | - | 1 | 20 | |||
| BC058210.1 | - | 9 | 1756 | |||
| BC141168.1 | - | 9 | 1316 | 9055780 | ||
| BC145480.1 | - | 10 | 1473 | |||

Slc2a6
Genomic Map