siRNA Details
| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178693.4 | NP_848808.1 | 4 | 336 | ||
| GenBank mRNA | AK039422.1 | - | 4 | 337 | ||
| AK039676.1 | - | 4 | 337 | |||
| AK043574.1 | - | 4 | 312 | |||
| AK050459.1 | - | 4 | 346 | |||
| AK163270.1 | - | 1105 | ||||
| BC139373.1 | - | 4 | 379 | 8862395 | ||
| BC139376.1 | - | 4 | 376 | 8862398 | ||
| BC145615.1 | - | 4 | 379 | |||

Coq4
Genomic Map