siRNA Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290503.1 | NP_001277432.1 | 12 | 2240 | ||
| NM_178694.4 | NP_848809.2 | 12 | 2279 | |||
| GenBank mRNA | AK155401.1 | - | 12 | 2187 | ||
| BC048924.1 | - | 12 | 2122 | 6312691 | ||
| BC057021.1 | - | 12 | 2184 | 6825937 | ||

Zer1
Genomic Map