siRNA Details
| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172660.4 | NP_766248.3 | 9 | 763 | ||
| GenBank mRNA | AK036945.1 | - | 7 | 1057 | ||
| AK044983.1 | - | 7 | 1357 | |||
| AK046148.1 | - | 8 | 958 | |||
| AK147162.1 | - | 9 | 753 | |||
| BC078441.1 | - | 7 | 921 | 5686308 | ||

D2Wsu81e
Genomic Map