siRNA Details
| Gene Symbol: BC005624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144885.2 | NP_659134.1 | 5, 6 | 419 | ||
| GenBank mRNA | AK158706.1 | - | 384 | |||
| AK163554.1 | - | 392 | ||||
| AK168109.1 | - | 5, 6 | 339 | |||
| BC005624.1 | - | 5, 6 | 364 | 3962182 | ||

BC005624
Genomic Map