siRNA Details
| Gene Symbol: Ppapdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145521.3 | NP_663496.1 | 1 | 500 | ||
| GenBank mRNA | AK052883.1 | - | 1 | 501 | ||
| AK134638.1 | - | 1 | 624 | |||
| BC016136.1 | - | 1 | 431 | 4910887 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc3| Gene Symbol: Ppapdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145521.3 | NP_663496.1 | 1 | 500 | ||
| GenBank mRNA | AK052883.1 | - | 1 | 501 | ||
| AK134638.1 | - | 1 | 624 | |||
| BC016136.1 | - | 1 | 431 | 4910887 | ||