siRNA Details
| Gene Symbol: Prrc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159634.1 | NP_001153106.1 | 25 | 5938 | ||
| NM_172661.3 | NP_766249.2 | 24 | 3880 | |||
| GenBank mRNA | AK052511.1 | - | 22 | 3463 | ||
| AK122300.1 | - | 10 | 1797 | |||
| AK147271.1 | - | 25 | 5938 | |||
| AK147677.1 | - | 25 | 5938 | |||
| BC055116.1 | - | 24 | 3871 | 6329995 | ||

Prrc2b
Genomic Map