siRNA Details
| Gene Symbol: Zp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011775.6 | NP_035905.1 | 7 | 684 | ||
| GenBank mRNA | AK135903.1 | - | 7 | 684 | ||
| AK138699.1 | - | 7 | 685 | |||
| BC071183.1 | - | 7 | 666 | 30473497 | ||
| M34148.1 | - | 7 | 684 | |||

Zp2
Genomic Map