siRNA Details
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172665.5 | NP_766253.2 | 2 | 377 | ||
| GenBank mRNA | AK038454.1 | - | 1 | 1892 | ||
| AK045337.1 | - | 1 | 1755 | |||
| AK046805.1 | - | 2 | 378 | |||
| AK085215.1 | - | 2 | 276 | |||
| AK153649.1 | - | 2 | 330 | |||
| AK163553.1 | - | 2 | 307 | |||
| BC027196.1 | - | 2 | 300 | 4458562 | ||

Pdk1
Genomic Map