siRNA Details
| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081260.2 | NP_001074729.1 | 6 | 3026 | ||
| GenBank mRNA | AK129436.1 | - | 2 | 1719 | ||
| BC025943.1 | - | 1 | 475 | |||
| BC058639.1 | - | 2 | 2078 | |||
| BC150947.1 | - | 6 | 2946 | 9087858 | ||

Tnks1bp1
Genomic Map