siRNA Details
Gene Symbol: Lrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145857.1 | NP_001139329.1 | 10 | 1238 | ||
NM_172668.3 | NP_766256.3 | 10 | 1238 | |||
GenBank mRNA | AF247637.1 | - | 10 | 1068 | ||
AK032360.1 | - | 10 | 1259 | |||
AK036406.1 | - | 2 | 96 | |||
AK051967.1 | - | 2 | 157 | |||
BC132240.1 | - | 10 | 1161 | 40130517 |
