siRNA Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172673.3 | NP_766261.2 | 7 | 799 | ||
| GenBank mRNA | AK030883.1 | - | 7 | 555 | ||
| AK044270.1 | - | 7 | 550 | |||
| AK048865.1 | - | 7 | 800 | |||
| AK054484.1 | - | 7 | 756 | |||
| BC038623.1 | - | 7 | 536 | |||
| BC057549.1 | - | 4 | 328 | 6416914 | ||
| BC062889.1 | - | 7 | 756 | 5698263 | ||

Frmd5
Genomic Map