siRNA Details
| Gene Symbol: Cstl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177655.3 | NP_808323.1 | 2 | 345 | ||
| GenBank mRNA | AK131797.1 | - | 2 | 346 | ||
| AK132807.1 | - | 2 | 348 | |||
| AY707915.1 | - | 2 | 345 | |||
| BC048646.1 | - | 2 | 345 | 6772392 | ||

Cstl1
Genomic Map