siRNA Details
| Gene Symbol: Tmem74bos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042610.1 | - | 2 | 372 | ||
| AK137371.1 | - | 2 | 373 | |||
| BC052486.1 | - | 2 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem74bos| Gene Symbol: Tmem74bos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042610.1 | - | 2 | 372 | ||
| AK137371.1 | - | 2 | 373 | |||
| BC052486.1 | - | 2 | 592 | |||