siRNA Details
Gene Symbol: Ralgapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291137.1 | NP_001278066.1 | 7 | 1317 | ||
NM_001291138.1 | NP_001278067.1 | 7 | 1317 | |||
GenBank mRNA | AK046067.1 | - | 7 | 1292 | ||
AK122470.1 | - | 1 | 100 | |||
BC138377.1 | - | 7 | 1219 | 8861397 | ||
BC145260.1 | - | 7 | 1219 | |||
CK638301.1 | - | 1 | 35 |
