siRNA Details
| Gene Symbol: Npepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213733.2 | NP_998898.1 | 7, 8 | 966 | ||
| GenBank mRNA | AK150313.1 | - | 7, 8 | 967 | ||
| AK156136.1 | - | 7, 8 | 967 | |||
| AK170509.1 | - | 7, 8 | 967 | |||
| BC023239.1 | - | 6, 7 | 644 | |||
| BC055448.1 | - | 6, 7 | 641 | |||
| BC069933.1 | - | 7, 8 | 961 | 4008091 | ||
| BC080707.1 | - | 7, 8 | 839 | |||

Npepl1
Genomic Map