siRNA Details
| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145919.1 | NP_001139391.1 | 4 | 1541 | ||
| NM_172677.3 | NP_766265.3 | 5 | 1574 | |||
| GenBank mRNA | AK039176.1 | - | 5 | 1637 | ||
| AK053736.1 | - | 5 | 1609 | |||
| AK078922.1 | - | 1 | 309 | |||
| AK137245.1 | - | 5 | 1521 | |||
| BC042627.1 | - | 1 | 1091 | |||
| BC052631.1 | - | 1 | 891 | |||
| BC057158.1 | - | 5 | 1601 | 5348717 | ||
| BC067040.1 | - | 4 | 1512 | 6853196 | ||
| BC067042.1 | - | 5 | 1507 | 6856248 | ||

Ythdf3
Genomic Map