siRNA Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145919.1 | NP_001139391.1 | 3 | 430 | ||
NM_172677.3 | NP_766265.3 | 3 | 430 | |||
GenBank mRNA | AK039176.1 | - | 4 | 526 | ||
AK053736.1 | - | 4 | 498 | |||
AK137245.1 | - | 3 | 377 | |||
BC057158.1 | - | 4 | 490 | 5348717 | ||
BC067040.1 | - | 3 | 401 | 6853196 | ||
BC067042.1 | - | 3 | 363 | 6856248 |
