siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 81 | 14340 | ||
GenBank mRNA | AK041642.1 | - | 8 | 1424 | ||
AK051829.1 | - | 1 | 1721 | |||
AK052702.1 | - | 13 | 2158 | |||
AK134811.1 | - | 2 | 310 | |||
AK173100.1 | - | 25 | 3975 | |||
BC024954.1 | - | 3 | 454 | |||
BC027125.1 | - | 8 | 1372 | |||
BC079623.1 | - | 22 | 3710 |
