siRNA Details
| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172679.2 | NP_766267.2 | 26 | 3497 | ||
| GenBank mRNA | AK030086.1 | - | 26 | 3483 | ||
| AK220265.1 | - | 26 | 3538 | |||
| BC034738.1 | - | 18 | 2346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932438A13Rik| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172679.2 | NP_766267.2 | 26 | 3497 | ||
| GenBank mRNA | AK030086.1 | - | 26 | 3483 | ||
| AK220265.1 | - | 26 | 3538 | |||
| BC034738.1 | - | 18 | 2346 | |||