siRNA Details
| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172679.2 | NP_766267.2 | 53 | 9595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932438A13Rik| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172679.2 | NP_766267.2 | 53 | 9595 | ||