siRNA Details
| Gene Symbol: Clrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153384.3 | NP_700433.1 | 4 | 713 | ||
| NM_153385.3 | NP_700434.1 | 3 | 659 | |||
| NM_153386.3 | NP_700435.1 | 2 | 479 | |||
| GenBank mRNA | AF495718.1 | - | 4 | 713 | ||
| AF495719.1 | - | 3 | 659 | |||
| AF495720.1 | - | 2 | 477 | |||
| AK037267.1 | - | 4 | 377 | |||
| AK041623.1 | - | 2 | 415 | |||
| AK042523.1 | - | 2 | 415 | |||
| BC132254.1 | - | 2 | 461 | 40130531 | ||
| BC132256.1 | - | 2 | 461 | 40130533 | ||
| BC145202.1 | - | 3 | 641 | 9053753 | ||

Clrn1
Genomic Map