siRNA Details
| Gene Symbol: Gpr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177346.4 | NP_796320.2 | 4 | 2667 | ||
| GenBank mRNA | AK035931.1 | - | 4 | 1230 | ||
| AK048373.1 | - | 1390 | ||||
| AK136826.1 | - | 4 | 2668 | |||
| AK166029.1 | - | 1495 | ||||
| BC119599.2 | - | 4 | 1331 | |||
More Information
Set Membership: |
|

Gpr149
Genomic Map