siRNA Details
| Gene Symbol: Ints3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145540.3 | NP_663515.2 | 4 | 1061 | ||
| NM_178876.3 | NP_849207.2 | 4 | 1061 | |||
| GenBank mRNA | AK033296.1 | - | 4 | 728 | ||
| AK040147.1 | - | 4 | 1051 | |||
| AK146699.1 | - | 4 | 955 | |||
| AK147865.1 | - | 4 | 1040 | |||
| AK188601.1 | - | 1 | 54 | |||
| BC055344.1 | - | 4 | 596 | 6827687 | ||

Ints3
Genomic Map