siRNA Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033302.2 | NP_001028474.1 | 6 | 1151 | ||
| GenBank mRNA | AK164001.1 | - | 6 | 1151 | ||
| BC132471.1 | - | 5 | 1047 | 40130748 | ||
| BC138510.1 | - | 5 | 1047 | 8861530 | ||

Ciart
Genomic Map