siRNA Details
| Gene Symbol: BC028528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153513.2 | NP_705733.2 | 3 | 163 | ||
| GenBank mRNA | AF534879.1 | - | 3 | 163 | ||
| BC028528.1 | - | 3 | 181 | 1366537 | ||
| Gene Symbol: C920021L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044447.1 | - | 4 | 1512 | ||

BC028528
Genomic Map