siRNA Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025613.1 | NP_001020784.1 | 9 | 1279 | ||
| NM_001025614.1 | NP_001020785.1 | 8 | 1221 | |||
| GenBank mRNA | AK044701.1 | - | 9 | 1280 | ||
| BC009124.1 | - | 3 | 188 | |||
| BC037040.1 | - | 11 | 1265 | |||
| BC141397.1 | - | 8 | 1111 | 9056010 | ||
| BC141398.1 | - | 8 | 1111 | 9056011 | ||

Otud7b
Genomic Map