siRNA Details
| Gene Symbol: Ak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081277.1 | NP_001074746.1 | 2 | 393 | ||
| GenBank mRNA | AK053807.1 | - | 2 | 393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak5| Gene Symbol: Ak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081277.1 | NP_001074746.1 | 2 | 393 | ||
| GenBank mRNA | AK053807.1 | - | 2 | 393 | ||